missing translation for 'onlineSavingsMsg'
Learn More

Mitapivat, MedChemExpress

Product Code. 16488007
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488007

Brand: Medchem Express HY12689

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mitapivat (AG-348) is a potent, orally active, and allosteric activator of pyruvate kinase with an AC50 of 20 nM. Mitapivat (AG-348) increases enzymatic activity, protein stability, and ATP levels over a broad range of PKLR genotypes. Mitapivat (AG-348) has the potential for restoring the glycolytic pathway activity with PK deficiency.

Mitapivat (AG-348) is a potent, orally active, and allosteric activator of pyruvate kinase with an AC50 of 20 nM. Mitapivat (AG-348) increases enzymatic activity, protein stability, and ATP levels over a broad range of PKLR genotypes. Mitapivat (AG-348) has the potential for restoring the glycolytic pathway activity with PK deficiency.

Specifications

Chemical Name or Material Mitapivat
CAS 1260075-17-9
Color White
Purity Grade Notes Research
Molecular Formula C24H26N4O3S
Quantity 5 mg
Synonym AG-348
Solubility Information DMSO : 20.83 mg/mL (46.23 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=C2N=CC=CC2=CC=C1)(NC3=CC=C(C(N4CCN(CC5CC5)CC4)=O)C=C3)=O
Molecular Weight (g/mol) 450.55
Formula Weight 450.55
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.