missing translation for 'onlineSavingsMsg'
Learn More

MIST1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18427065 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427065 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427065 Supplier Novus Biologicals Supplier No. NBP282007AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MIST1 Polyclonal antibody specifically detects MIST1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIST1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias basic helix-loop-helix domain containing, class B, 8, basic helix-loop-helix family, member a15, bHLHa15, bHLHb8, class A basic helix-loop-helix protein 15, Class B basic helix-loop-helix protein 8, class II bHLH protein MIST1, MIST1MIST-1, Muscle, intestine and stomach expression 1
Host Species Rabbit
Immunogen MIST1 antibody was raised against a 18 amino acid peptide near the center of human MIST1. The immunogen is located within amino acids 60 - 110 of MIST1. Amino Acid Squence: SIQRRLESNERERQRMHK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168620
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.