missing translation for 'onlineSavingsMsg'
Learn More

Misoprostol Acid (10 mg in 1 mL Methyl Acetate), TRC

Product Code. 30009807
Change view
Click to view available options
Quantity:
0.1 mL
0.25 mL
1 mL
Unit Size:
0.1mL
0.25mL
1mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009807 0.1 mL 0.1mL
30004065 0.25 mL 0.25mL
30004788 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009807 Supplier Toronto Research Chemicals Supplier No. TRCM3687600.1ML
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112137-89-0
Molecular Formula C21 H36 O5
Molecular Weight (g/mol) 368.51
Synonym Misoprostol Acid
IUPAC Name 7-[(1R,2R,3R)-3-hydroxy-2-[(E)-4-hydroxy-4-methyloct-1-enyl]-5-oxocyclopentyl]heptanoic acid
SMILES CCCCC(C)(O)C\C=C\[C@H]1[C@H](O)CC(=O)[C@@H]1CCCCCCC(=O)O

Specifications

Quantity 0.1 mL
InChI Formula InChI=1S/C21H36O5/c1-3-4-13-21(2,26)14-9-11-17-16(18(22)15-19(17)23)10-7-5-6-8-12-20(24)25/h9,11,16-17,19,23,26H,3-8,10,12-15H2,1-2H3,(H,24,25)/b11-9+/t16-,17-,19-,21?/m1/s1
Formula Weight 368.2563
Concentration or Composition (by Analyte or Components) Misoprostol Acid 10mg/mL
Chemical Name or Material Misoprostol Acid (10 mg in 1 mL Methyl Acetate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.