missing translation for 'onlineSavingsMsg'
Learn More

Misoprostol, 1% in cellulose, TRC

Product Code. 30179462
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179462 1 mg 1mg
30179186 10 mg 10mg
30179124 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179462 Supplier Toronto Research Chemicals Supplier No. TRCM3687501MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59122-46-2
Molecular Formula C22 H38 O5
Molecular Weight (g/mol) 382.53
Synonym Misoprostol, Mixture of methyl 7-[(1RS,2RS,3RS)-3-hydroxy-2-[(1E,4RS)-4-hydroxy-4-methyloct-1-enyl]-5-oxocyclopentyl]heptanoate and methyl 7-[(1RS,2RS,3RS)-3-hydroxy-2-[(1E,4SR)-4-hydroxy-4-methyloct-1-enyl]-5-oxocyclopentyl]heptanoate, (11α,13E)-(+/-)-11,16-Dihydroxy-16-methyl-9-oxoprost-13-en-1-oic Acid Methyl Ester, Cytotec, Misogon, Misoprostil, SC 29333;
IUPAC Name methyl 7-[(1R,2R,3R)-3-hydroxy-2-[(E)-4-hydroxy-4-methyloct-1-enyl]-5-oxocyclopentyl]heptanoate
SMILES CCCCC(C)(CC=CC1C(CC(=O)C1CCCCCCC(=O)OC)O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C22H38O5/c1-4-5-14-22(2,26)15-10-12-18-17(19(23)16-20(18)24)11-8-6-7-9-13-21(25)27-3/h10,12,17-18,20,24,26H,4-9,11,13-16H2,1-3H3/b12-10+/t17-,18-,20-,22?/m1/s1
Formula Weight 382.2719
Concentration or Composition (by Analyte or Components) Misoprostol 1%
Chemical Name or Material Misoprostol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.