missing translation for 'onlineSavingsMsg'
Learn More

Misonidazole, TRC

Product Code. 18073210
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18073210 100 mg 100mg
18040520 1 g 1g
17909949 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18073210 Supplier Toronto Research Chemicals Supplier No. TRCM368700100MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13551-87-6
Molecular Formula C7 H11 N3 O4
Molecular Weight (g/mol) 201.18
Synonym α-(Methoxymethyl)-2-nitro-1H-imidazole-1-ethanol, α-(Methoxymethyl)-2-nitroimidazole-1-ethanol, 1-(2-Hydroxy-3-methoxypropyl)-2-nitroimidazole, 1-(2-Nitro-1-imidazolyl)-3-methoxy-2-propanol, 2-Nitro-1H-imidazole-1-(3-methoxy-2-propanol), NSC 261037, Racemic Misonidazole, Ro 7-0582, SR 1354, SRI 1354
IUPAC Name 1-methoxy-3-(2-nitroimidazol-1-yl)propan-2-ol
SMILES COCC(O)Cn1ccnc1[N+](=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C7H11N3O4/c1-14-5-6(11)4-9-3-2-8-7(9)10(12)13/h2-3,6,11H,4-5H2,1H3
Formula Weight 201.075
Chemical Name or Material Misonidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.