missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Miscellaneous Microtome Section Waste Tray with Arm Rest, For HM 310, HM 315, HM325, and HM 335

Product Code. 12063089 Shop All Epredia Products
Click to view available options
Product Type:
Foot Pedal
Large-field Magnifier
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12063089

Brand: Epredia™ 640280

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Support and expand the functionality of Epredia™ 200 and 300 series of microtomes with Epredia™ Miscellaneous Microtome Accessories.

  • Interlock plug replaces foot pedal connector
  • Storage unit can be removed and used separately; tray for specimens can be cooled by changeable inserts
  • Large-field magnifier attaches to facilitate knife alignment and approach for sectioning; magnification 2.5X
TRUSTED_SUSTAINABILITY

Specifications

Product Type Foot Pedal
For Use With (Equipment) Epredia 200/300 Series Microtomes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.