missing translation for 'onlineSavingsMsg'
Learn More

MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 μg/mL in 2% HNO3, VHG™

Product Code. 30003158
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
30003158 500 mL 500mL
30003103 100 mL 100mL
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30003158

Brand: VHG Labs VHGMISA5500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

MISA Standard 5: Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb @ 100 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Holmium/Erbium/Samarium/Thulium/Caesium/Terbium/Scandium/Dysprosium/Europium/Neodymium/Ytterbium/Lanthanum/Praseodymium/Thorium/Yttrium/Gadolinium/Lutetium
CAS 7440-60-0, 7440-52-0, 7440-19-9, 7440-30-4, 7440-46-2, 7440-27-9
Methods ICP/ICP-MS
Analyte or Component Names Holmium/Erbium/Samarium/Thulium/Caesium/Terbium/Scandium/Dysprosium/Europium/Neodymium/Ytterbium/Lanthanum/Praseodymium/Thorium/Yttrium/Gadolinium/Lutetium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Holmium 99.88 μg/mL/Erbium 99.94 μg/mL/Samarium 99.95 μg/mL/Thulium 99.96 μg/mL/Caesium 99.97 μg/mL/Terbium 99.97 μg/mL/Scandium 99.98 μg/mL/Dysprosium 99.99 μg/mL/Europium 100 μg/mL/Neodymium 100 μg/mL/Ytterbium 100 μg/mL/Lanthanum 100.1 μg/mL/Praseodymium 100.1 μg/mL/Thorium 100.1 μg/mL/Yttrium 100.1 μg/mL/Gadolinium 100.2 μg/mL/Lutetium 100.2 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.