missing translation for 'onlineSavingsMsg'
Learn More

MISA Standard 4: As, Be, Bi, Hg, Re, S, Se @ 100 μg/mL in 10% HNO3, VHG™

Product Code. 30003126
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003126 100 mL 100mL
30002978 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30003126 Supplier VHG Labs Supplier No. VHGMISA4100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

MISA Standard 4: As, Be, Bi, Hg, Re, S, Se @ 100 μg/mL in 10% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mercury/Rhenium/Arsenic/Beryllium/Bismuth/Selenium/Sulfur
CAS 7439-97-6, 7440-15-5, 7440-38-2, 7440-41-7, 7440-69-9, 7782-49-2
Methods ICP/ICP-MS
Analyte or Component Names Mercury/Rhenium/Arsenic/Beryllium/Bismuth/Selenium/Sulfur
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Mercury 99.95 μg/mL/Rhenium 99.98 μg/mL/Arsenic 99.99 μg/mL/Beryllium 100 μg/mL/Bismuth 100 μg/mL/Selenium 100 μg/mL/Sulfur 100.1 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.