missing translation for 'onlineSavingsMsg'
Learn More

MISA Standard 1: Ge, Hf, Mo, Nb, Si, Ta, Ti, W, Zr @ 100 μg/mL in H2O/tr. HF, VHG™

Product Code. 17971336
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971336 500 mL 500mL
17972016 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971336 Supplier VHG Labs Supplier No. VHGMISA1500
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

MISA Standard 1: Ge, Hf, Mo, Nb, Si, Ta, Ti, W, Zr @ 100 μg/mL in H2O/tr. HF
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Germanium/Hafnium/Molybdenum/Niobium/Tantalum/Titanium/Tungsten/Zirconium
CAS 7440-56-4, 7440-58-6, 7439-98-7, 3-1-7660, 7440-25-7, 7440-32-6
Methods ICP/ICP-MS
Analyte or Component Names Germanium/Hafnium/Molybdenum/Niobium/Tantalum/Titanium/Tungsten/Zirconium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HCl
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Germanium 100 μg/mL/Hafnium 100 μg/mL/Molybdenum 100 μg/mL/Niobium 100 μg/mL/Tantalum 100 μg/mL/Titanium 100 μg/mL/Tungsten 100 μg/mL/Zirconium 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.