missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIS18A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554238
This item is not returnable.
View return policy
Description
Immunogen sequence: MAGVRSLRCS RGCAGGCECG DKGKCSDSSL LGKRLSEDSS RHQLLQKWAS MWSSMSEDAS VADMERAQLE EEAAAAEERP LVF Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 57%.
C21orf45, also named as MIS18A, C21orf46 and FASP1, is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. C21orf45 has dimer (~52 kDa) and heterodimer(~105 kDa) forms.
Specifications
| MIS18A | |
| Polyclonal | |
| Unconjugated | |
| MIS18A | |
| 2610039C10Rik; 2810018N07Rik; B28; C21orf45; C21orf46; FAPP1-associated protein 1; FASP1; hMis18alpha; MIS18 kinetochore protein A; MIS18 kinetochore protein homolog A; MIS18 kinetochore protein homolog A (S. pombe); Mis18a; MIS18alpha; protein Mis18-alpha; RGD1310778 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54069 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NYP9 | |
| MIS18A | |
| Recombinant protein corresponding to Human MIS18A. Recombinant protein control fragment (Product #RP-91767). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction