missing translation for 'onlineSavingsMsg'
Learn More

MIS/AMH Antibody (805531) [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30735119 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735119 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735119 Supplier Novus Biologicals Supplier No. FAB17371J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIS/AMH Monoclonal antibody specifically detects MIS/AMH in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIS/AMH
Applications Immunocytochemistry
Classification Monoclonal
Clone 805531
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MIS/AMH, Leu19-Gln450 (predicted), Accession # P03971
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.