missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIS/AMH Antibody (003), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90330CL1
This item is not returnable.
View return policy
Description
MIS/AMH Monoclonal antibody specifically detects MIS/AMH in Human samples. It is validated for ELISA,Sandwich ELISA Detection
Specifications
| MIS/AMH | |
| Monoclonal | |
| CoraFluor 1 | |
| MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human MIS/AMH (Accession#: NP_000470.2; Leu25-Arg560). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA Detection | |
| 003 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 268 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction