missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine N-Oxide, LoGiCal

Product Code. 30460919
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460919 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460919 Supplier LoGiCal Supplier No. LGCFOR1005.02
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mirtazapine N-Oxide
CAS 155172-12-6
Analyte or Component Names Mirtazapine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O
Certifications/Compliance ISO 9001
Synonym Mirtazapine Imp. A (EP), Mirtazapine USP Related Compound B, Mirtazapine USP RC B, (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-Oxide, Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepine, 1,2,3,4,10,14b-hexahydro-2-methyl-, 2-oxide, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide, Mirtazapine Impurity A
InChI Formula InChI=1S/C17H19N3O/c1-20(21)10-9-19-16(12-20)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)19/h2-8,16H,9-12H2,1H3
Solution Type Neat
SMILES C[N+]1([O-])CCN2C(C1)c3ccccc3Cc4cccnc24
IUPAC Name (14bRS)-2-Methyl-1,2,3,4,10-14b-hexahydropyrazino[2,1-a]pyrido[2,3-c][2]benzazepine 2-oxide
Industry Type Clinical Testing
Molecular Weight (g/mol) 281.35
Quantity 10 mg
Formula Weight 281.1528 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.