missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine, MedChemExpress

Product Code. p-200100052
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. Quantity unitSize
16428256 100 mg 100mg
16402116 500 mg 500mg
16401416 200 mg 200mg
3 options
This item is not returnable. View return policy

Product Code. 16428256

Brand: Medchem Express HYB0352

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3 and 6.95, respectively.

Mirtazapine (Org3770) is a potent and orally active noradrenergic and specific serotonergic antidepressant (NaSSA) agent. Mirtazapine is also a 5-HT2, 5-HT3, histamine H1 receptor and α2-adrenoceptor antagonist with pKi values of 8.05, 8.1, 9.3 and 6.95, respectively.

Specifications

Chemical Name or Material Mirtazapine
CAS 85650-52-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H19N3
Quantity 100 mg
Synonym Org3770 6-Azamianserin
Solubility Information DMSO : 50 mg/mL (188.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.52 mM; Need ultrasonic)
SMILES CN1CC2C3=CC=CC=C3CC4=CC=CN=C4N2CC1
Molecular Weight (g/mol) 265.35
Formula Weight 265.35
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.