missing translation for 'onlineSavingsMsg'
Learn More

Mirtazapine, Lipomed

Product Code. 30462531
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462531 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462531 Supplier Lipomed Supplier No. LPMMRT1166FB50
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Mirtazapine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mirtazapine
CAS 85650-52-8
Analyte or Component Names Mirtazapine
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3
Synonym 1,2,3,4,10,14 b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2 ]benzazepine, (+/-)-1,2,3,4,10,14 b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2 ]benzazepine, 6-Azamianserin, Mepirzapin, Mepirzepine, Mirtazapine, Mirtazepine, Mirtazipine, Org 3770, Promyrtil, Remergil, Remergon, Remeron, Rexer, Zispin
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCN2C(C1)c3ccccc3Cc4cccnc24
IUPAC Name 1,2,3,4,10,14 b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2 ]benzazepine
Industry Type Pharmaceutical
Molecular Weight (g/mol) 265.35
Quantity 50 mg
Formula Weight 265.1579 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.