Learn More
Edulab Mirror, Convex - Pack of 10
Product Code. 30280303
Change view
Click to view available options
Product Type:
Convex Mirror
Unit Size:
Pack of 10
Product Code. Product Type Unit Size
30280303 Convex Mirror Pack of 10
1 options
Product Code. 30280303 Supplier Edulab Supplier No. PH0526D
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Convex mirror designed for classroom optics experiments and practical demonstrations.
  • 50 mm diameter convex mirror for practical reflection experiments
  • Optically worked to provide clear and accurate reflections
  • Focal length of 150 mm for standard classroom use
  • Silvered back with protective coating for durability
Specifications
Product Type Convex Mirror