missing translation for 'onlineSavingsMsg'
Learn More

Mirogabalin, MedChemExpress

Product Code. p-200099286
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483316

Brand: Medchem Express HY12650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS.

Mirogabalin (DS-5565) is a novel, preferentially selective α2δ-1 ligand characterized by high potency and selectivity to the α2δ-1 subunit of voltage-sensitive calcium channel complexes in the CNS.

Specifications

Chemical Name or Material Mirogabalin
Physical Form Solid
CAS 1138245-13-2
Color Light Brown
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C12H19NO2
For Use With (Application) Neuroscience-Neuromodulation
Synonym DS5565
Solubility Information H2O : 7.71 mg/mL (36.84 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=C(O)C[C@]1(CN)[C@]2([H])C=C(CC)C[C@]2([H])C1
Molecular Weight (g/mol) 209.28
Formula Weight 209.28
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.