missing translation for 'onlineSavingsMsg'
Learn More

Mirogabalin besylate, MedChemExpress

Product Code. p-200098755
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16474168

Brand: Medchem Express HY108006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mirogabalin besylate is a selective and orally available ligand for the α2δ subunit of voltage-gated calcium channels, with Kds of 13.5 nM, 22.7 nM, 27 nM, and 47.6 nM for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2, respectively.

Mirogabalin besylate is a selective and orally available ligand for the α2δ subunit of voltage-gated calcium channels, with Kds of 13.5 nM, 22.7 nM, 27 nM, and 47.6 nM for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2, respectively.

Specifications

Chemical Name or Material Mirogabalin besylate
CAS 1138245-21-2
Color White
Purity Grade Notes Research
Molecular Formula C18H25NO5S
Quantity 10 mg
Synonym DS 5565 besylate
Solubility Information DMSO : 125 mg/mL (340.17 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(O)=O.OC(C[C@@]2([C@@]3([H])[C@@](CC(CC)=C3)([H])C2)CN)=O
Molecular Weight (g/mol) 367.46
Formula Weight 367.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.