missing translation for 'onlineSavingsMsg'
Learn More

Mirogabalin besylate, MedChemExpress

Product Code. 16474168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474168 10 mg 10mg
16460517 50 mg 50mg
16436877 5 mg 5mg
16405136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16474168 Supplier Medchem Express Supplier No. HY108006
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mirogabalin besylate is a selective and orally available ligand for the α2δ subunit of voltage-gated calcium channels, with Kds of 13.5 nM, 22.7 nM, 27 nM, and 47.6 nM for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2, respectively.

Mirogabalin besylate is a selective and orally available ligand for the α2δ subunit of voltage-gated calcium channels, with Kds of 13.5 nM, 22.7 nM, 27 nM, and 47.6 nM for human α2δ-1, human α2δ-2, rat α2δ-1, and rat α2δ-2, respectively.

Specifications

Chemical Name or Material Mirogabalin besylate
CAS 1138245-21-2
Color White
Purity Grade Notes Research
Molecular Formula C18H25NO5S
Quantity 10 mg
Synonym DS 5565 besylate
Solubility Information DMSO : 125 mg/mL (340.17 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1)(O)=O.OC(C[C@@]2([C@@]3([H])[C@@](CC(CC)=C3)([H])C2)CN)=O
Molecular Weight (g/mol) 367.46
Formula Weight 367.46
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.