missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Mirin

Product Code. 18036022
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18036022 10 mg 10mg
18004372 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18036022 Supplier Tocris Bioscience™ Supplier No. 3190/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MRN-ATM pathway inhibitor

Mre11-Rad50-Nbs1 (MRN)-ATM pathway inhibitor that blocks the 3' and 5' exonuclease activity associated with Mre11. Prevents ATM activation in response to double strand breaks (IC50 = 12 μM) and induces G2 cell cycle arrest. Also blocks homology-directed repair in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198097-97-0
Molecular Formula C10H8N2O2S

Specifications

Quantity 10 mg
Formula Weight 220.25
Purity >99%
Molecular Weight (g/mol) 220.246
Chemical Name or Material Mirin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.