missing translation for 'onlineSavingsMsg'
Learn More

Mirdametinib, MedChemExpress

Product Code. 16454806
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454806 100 mg 100mg
16430127 50 mg 50mg
16406477 5 mg 5mg
16403748 10 mg 10mg
16402846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16454806 Supplier Medchem Express Supplier No. HY10254

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mirdametinib (PD0325901) is an orally active, selective and non-ATP-competitive MEK inhibitor with an IC50 of 0.33 nM. Mirdametinib exhibits a Kiapp of 1 nM against activated MEK1 and MEK2. Mirdametinib suppresses the expression of p-ERK1/2 and induces apoptosis. Mirdametinib has anti-cancer activity for a broad spectrum of human tumor xenografts.

Mirdametinib (PD0325901) is an orally active, selective and non-ATP-competitive MEK inhibitor with an IC50 of 0.33 nM. Mirdametinib exhibits a Kiapp of 1 nM against activated MEK1 and MEK2. Mirdametinib suppresses the expression of p-ERK1/2 and induces apoptosis. Mirdametinib has anti-cancer activity for a broad spectrum of human tumor xenografts.

Specifications

Chemical Name or Material Mirdametinib
CAS 391210-10-9
Color White
Purity Grade Notes Research
Molecular Formula C16H14F3IN2O4
Quantity 100 mg
Synonym PD0325901 PD325901
Solubility Information DMSO : ≥ 56 mg/mL (116.14 mM)
SMILES O=C(C1=CC=C(C(F)=C1NC2=CC=C(I)C=C2F)F)NOC[C@H](O)CO
Molecular Weight (g/mol) 482.19
Formula Weight 482.19
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.