Learn More
PeproTech MIP-3 alpha Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | MIP-3 alpha |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17893613
|
PeproTech
500-P95ABT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17823393
|
PeproTech
500-P95ABT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17843683
|
Invitrogen
500-P95ABT-1MG |
2 x 500 μg | |||||||
|
30054662
|
PeproTech
500-P95ABT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKNM. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-3alpha (CCL20). Anti-Human MIP-3alpha (CCL20)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human MIP-3alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human MIP-3alpha (500-P95A) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MIP-3alpha. Western Blot: To detect Human MIP-3alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human MIP-3alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P95ABT-1MG will be provided as 2 x 500 μg
Macrophage Inflammatory Protein-3 alpha (MIP-3alpha), also called CCL20, is expressed in the liver, lungs, lymph nodes and peripheral blood lymphocytes. MIP-3alpha is strongly up regulated by inflammatory signals, and down regulated by the anti-inflammatory cytokine IL-10. MIP3alpha signals through the G protein-coupled receptor, CCR6, and acts as a chemoattractant to lymphocytes and dendritic cells.Specifications
| MIP-3 alpha | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| P78556 | |
| 6364 | |
| E.coli-derived, 8.0 kDa Recombinant Human MIP-3alpha (CCL20). | |
| Antigen affinity chromatography | |
| CCL20 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| beta chemokine exodus-1; beta-chemokine exodus-1; CC chemokine LARC; CC chemokine ST38; C-C motif chemokine 20; C-C motif chemokine ligand 20; Ccl20; CCL20(1-64); CCL20(1-67); CCL20(2-70); chemokine (C-C motif) ligand 20; CKb4; Exodus; exodus-1; LARC; Liver and activation-regulated chemokine; macrophage inflammatory protein 3 alpha; MIP 3 a; MIP 3 α; MIP 3alpha; MIP 3α; MIP3 a; MIP3 alpha; MIP3 α; MIP-3[a]; MIP3A; MIP-3A; MIP-3-alpha; M-MIP-3-alpha; SCYA20; small inducible cytokine subfamily A (Cys-Cys), member 20; small inducible cytokine subfamily A20; small-inducible cytokine A20; St38 | |
| CCL20 | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.