missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIP-1 delta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568468
This item is not returnable.
View return policy
Description
Predicted to react with Human samples.
This gene is located in a cluster of similar genes in the same region of chromosome 17. These genes encode CC cytokines, which are secreted proteins characterized by two adjacent cysteines. The product of this gene is chemotactic for T cells and monocytes, and acts through C-C chemokine receptor type 1 (CCR1). The proprotein is further processed into numerous smaller functional peptides. Naturally-occurring readthrough transcripts occur from this gene into the downstream gene, CCL14 (chemokine (C-C motif) ligand 14).
Specifications
| MIP-1 delta | |
| Polyclonal | |
| Unconjugated | |
| CCL15 | |
| C-C motif chemokine 15; C-C motif chemokine ligand 15; CCL15; CCL15(22-92); CCL15(25-92); CCL15(29-92); chemokine (C-C motif) ligand 15; chemokine CC-2; HCC-2; HCC-2Lkn-1; HMRP-2B; leukotactin 1; Leukotactin-1; LKN1; LKN-1; Macrophage inflammatory protein 5; MIP-1 delta; MIP-1D; MIP5; MIP-5; MIP-5MIP-1d; Mrp-2b; NCC3; NCC-3; NCC-3MIP-1D; NCC3MRP-2B; new CC chemokine 3; SCYA15; SCYA15member 15; SCYL3; small inducible cytokine subfamily A (Cys-Cys), member 15; small-inducible cytokine A15; SY15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6359 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16663 | |
| CCL15 | |
| Synthesized peptide derived from human MIP-1 delta [aa 51-100]. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction