missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NLRP3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142555
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_004886.3; NP_899632.1; NP_001120933.1; NP_001120934.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a pyrin-like protein containing a pyrin domain, a nucleotide-binding site domain, and a leucine-rich repeat motif. This protein interacts with the apoptosis-associated speck-like protein PYCARD/ASC, which contains a caspase recruitment domain, and is a member of the NALP3 inflammasome complex. This complex functions as an upstream activator of NF-kappaB signaling, and it plays a role in the regulation of inflammation, the immune response, and apoptosis. Mutations in this gene are associated with familial cold autoinflammatory syndrome, and neonatal-onset multisystem inflammatory disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. Alternative 5' UTR structures are suggested by available data; however, insufficient evidence is available to determine if all of the represented 5' UTR splice patterns are biologically valid.
Specifications
| NLRP3 | |
| Polyclonal | |
| Unconjugated | |
| NLRP3 | |
| AGTAVPRL; AII; AII/AVP; Angiotensin/vasopressin receptor AII/AVP-like; AVP; C1orf7; caterpiller protein 1.1; Cias1; CLR1.1; cold autoinflammatory syndrome 1 homolog; cold autoinflammatory syndrome 1 protein; cold autoinflammatory syndrome 1 protein homolog; Cold-induced autoinflammatory syndrome 1 protein; cryopyrin; FCAS; FCAS1; FCU; FLJ95925; mast cell maturation inducible protein 1; mast cell maturation-associated-inducible protein 1; Mmig1; MWS; NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3; NACHT, LRR and PYD containing protein 3; NACHT, LRR and PYD domains-containing protein 3; NACHT/LRR/pyrin domain-containing protein 3; NALP3; NLR family pyrin domain containing 3; NLR family, pyrin domain containing 3; Nlrp3; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3; PYPAF1; PYRIN-containing APAF1-like protein 1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 114548 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96P20 | |
| NLRP3 | |
| Peptide with sequence RDLYEKAKRDEPK-C. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction