missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: PEAFYMIRFH SFYPWHTGRD YQQLCSQQDL AMLPWVREFN KFDLYTKCPD LPDVDKLRPY YQGLIDKYCP GI Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
MIOX (Myo-Inositol Oxygenase) is a Protein Coding gene. Among its related pathways are superpathway of inositol phosphate compounds and Porphyrin and chlorophyll metabolism. Gene Ontology (GO) annotations related to this gene include iron ion binding and oxidoreductase activity, acting on NAD(P)H.
Specifications
Specifications
| Antigen | MIOX |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MIOX |
| Gene Accession No. | Q9UGB7 |
| Gene Alias | 0610009I10Rik; AI314022; aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; aldehyde reductase-like 6; Aldrl6; C85427; Inositol oxygenase; kidney-specific protein 32; KSP32; MI oxygenase; Miox; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxidoreductase; renal-specific oxido-reductase; Rsor |
| Gene Symbols | MIOX |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
