missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35432-100ul
This item is not returnable.
View return policy
Description
MIOX Polyclonal antibody specifically detects MIOX in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| MIOX | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| aldehyde reductase (aldose reductase) like 6, Aldehyde reductase-like 6, ALDRL6, inositol oxygenase, Kidney-specific protein 32, KSP32, MGC90217, MI oxygenase, myo-inositol oxygenaseEC 1.13.99.1, Renal-specific oxidoreductase, RSOR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human MIOX (NP_060054.4).,, Sequence:, MKVTVGPDPSLVYRPDVDPEVAKDKASFRNYTSGPLLDRVFTTYKLMHTHQTVDFVRSKHAQFGGFSYKKMTVMEAVDLLDGLVDESDPDVDFPNSFHAFQTAEGIRKAHPDKDWFHLVGLLHDLGKVLALFGEPQWAVVGDTFPVGCRPQASVVFCDSTFQDNPDLQDPRYSTELGMYQPHCGLDRVLMSWGHDEYMYQVMKFNKFSLPPEAFYMIRFHSFYPWHTGRDYQQLCSQQDLAMLPWVREFNKFDLYTKCPDLPDVDKLRPYYQGLIDKYCPGILSW | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55586 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction