missing translation for 'onlineSavingsMsg'
Learn More

Minoxidil

Product Code. 15464319
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464319 25 g 25g
15444319 1 g 1g
15454319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15464319 Supplier Thermo Scientific Alfa Aesar Supplier No. J61803.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Minoxidil, CAS # 38304-91-5, is a comopund with vasodilating effects that works by opening adenosine triphosphate (ATP)-sensitive potassium channels. | CAS: 38304-91-5 | C9H15N5O | 209.253 g/mol

Minoxidil is used as antihypertensive, antialopecia agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Minoxidil is a piperidine that acts as a vasodilator when converted into its active metabolite, minoxidil sulphate
  • Minoxidil opens adenosine triphosphate (ATP)-sensitive potassium channels in the plasma membrane and relaxes smooth muscle in arteriole walls. Relaxation of smooth muscle within the arterioles leads to decreased resistance and lower blood pressure

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38304-91-5
Molecular Formula C9H15N5O
Molecular Weight (g/mol) 209.253
MDL Number MFCD00063409
InChI Key ZIMGGGWCDYVHOY-UHFFFAOYSA-N
Synonym minoxidil, rogaine, loniten, regaine, minoximen, theroxidil, tricoxidil, alopexil, alostil, lonolox
PubChem CID 4201
IUPAC Name 3-hydroxy-2-imino-6-piperidin-1-ylpyrimidin-4-amine
SMILES C1CCN(CC1)C2=NC(=N)N(C(=C2)N)O

Specifications

Melting Point 248°C to 265°C
Color White
Quantity 25 g
Merck Index 14,6203
Solubility Information Soluble in water (2.2mg/ml),100% ethanol (29mg/ml),propylene glycol,acetone,DMSO (6.5mg/ml),and methanol.
Formula Weight 209.25
Physical Form Crystalline Powder
Chemical Name or Material Minoxidil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.