missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Minoxidil

Product Code. 18732554
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18732554 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18732554 Supplier R&D Systems™ Supplier No. 0583/100
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Kir6 channel (KATP) opener

Antihypertensive. Antialopecia agent. Kir6 channel (KATP) activator.
TRUSTED_SUSTAINABILITY

Specifications

CAS 38304-91-5
Quantity 100 mg
Product Line Tocris
Molecular Formula C9H15N5O
Formula Weight 209.25
Purity >99%
Molecular Weight (g/mol) 209.253
Chemical Name or Material Minoxidil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.