missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human MINK1, GST Tag Recombinant Protein

Product Code. 15478533
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15478533

Brand: Thermo Scientific™ PR7198A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

MINK1 is a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N4C8
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 2mM DTT, 150mM NaCl, 0.02% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 50488
Molecular Weight (g/mol) 63.9 kDa
Name Human MINK1, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias B55; FLJ18426; FLJ38123; FLJ94103; GCK family kinase MiNK; hMINK; hMINKbeta; ISK; JLNS; JLNS2; LQT2/5; LQT5; Map4k6; MAPK/ERK kinase kinase kinase 6; MEK kinase kinase 6; MEKKK 6; MGC33114; MINK; Mink 1; Mink 2; MINK1; misshapen like kinase 1; misshapen/NIK-related kinase; Misshapen/NIKs-related kinase; misshapen-like kinase 1; misshapen-like kinase 1 (zebrafish); mitogen-activated protein kinase kinase kinase kinase 6; Yeast Sps1/Ste20-related kinase 2; Ysk2; ZC3
Common Name MINK1
Gene Symbol MINK1
KinaseFamily STE20 Kinase Family
Protein Length 1-320
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 1-320
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: STE Group
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.