missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MINK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533936
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Mouse, Rat, Elephant, Panda, Bovine, Bat, Horse, Pig (100%) Opossum (94%).
MINK1 is a serine/threonine kinase belonging to the germinal center kinase (GCK) family. The protein is structurally similar to the kinases that are related to NIK and may belong to a distinct subfamily of NIK-related kinases within the GCK family. Studies of the mouse homolog indicate an up-regulation of expression in the course of postnatal mouse cerebral development and activation of the cJun N-terminal kinase (JNK) and the p38 pathways.
Specifications
| MINK1 | |
| Polyclonal | |
| Unconjugated | |
| MINK1 | |
| B55; FLJ18426; FLJ38123; FLJ94103; GCK family kinase MiNK; hMINK; hMINKbeta; ISK; JLNS; JLNS2; LQT2/5; LQT5; Map4k6; MAPK/ERK kinase kinase kinase 6; MEK kinase kinase 6; MEKKK 6; MGC33114; MINK; Mink 1; Mink 2; MINK1; misshapen like kinase 1; misshapen/NIK-related kinase; Misshapen/NIKs-related kinase; misshapen-like kinase 1; misshapen-like kinase 1 (zebrafish); mitogen-activated protein kinase kinase kinase kinase 6; Yeast Sps1/Ste20-related kinase 2; Ysk2; ZC3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100061370, 100515409, 303259, 50488, 50932, 519558 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| F1LP90, Q8N4C8, Q9JM52 | |
| MINK1 | |
| Synthetic 18 amino acid peptide from internal region of human MINK1 / MAP4K6. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Monkey, Bovine, Pig, Horse, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction