missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Minisart™ PES Venting Filter

Sartorius Stedim Biotech™ Minisart™ PES Venting Filter is a premium quality, hydrophobic PES filter designed for venting and gas filtration.

Brand:  Sartorius Stedim Biotech™ 1757H----------Q

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 15688058

  • £1120.00 / Pack of 500
In Stock
to see stock.
Add to basket

Description

Description

  • Premium filters for fail-safe filtration
  • Can be used for sterile filtration
  • Suitable for venting or filtration of gases and air
  • Can be sterilized by gamma irradiation, autoclaving or EO
Specifications

Specifications

White
PES
Non-sterile
Polypropylene
26 mm
Male luer slip connection
0.2 μm
Venting and gas filtration
500/pk.
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers