missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Minimum Essential Medium Eagle, Liquid

Product Code. 11450415 Shop All MP Biomedicals Products
Click to view available options
With Additives:
Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate
Hanks’ Balanced Salt solution (HBSS), Sodium Bicarbonate
Sodium Bicarbonate, Earle's Balanced Salt Solution (EBSS)
Without Additives:
No Glutamine
No Glutamine, No Cysteine, No Cystine, No Methionine
Product Type:
Minimum Essential Medium Eagle's
Minimum Essential Medium Eagle's (modified)
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11450415

Brand: MP Biomedicals™ 091641454

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Incorporates higher amino acid concentrations for the cultivation of fastidious cells

MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Upon his attempts to cultivate normal mammalian fibroblasts and certain HeLa cell subtypes, it was revealed that the nutritional needs of these cell types could not be met by BME. Further studies led to the development of MEM from MP Biomedicals incorporating specific modifications such as higher amino acid concentrations for the cultivation of fastidious cells.

  • MEM from MP Biomedicals may be used to support the growth of cells in monolayers, in suspension and wide variety of other cell types with proper supplementation.
  • Available with either Earle's or Hank's salts.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Minimum Essential Medium Eagle's (modified)
Sterility Sterile
With Additives Sodium Bicarbonate, Earle's Balanced Salt Solution (EBSS)
Without Additives No Glutamine, No Cysteine, No Cystine, No Methionine
Quantity 500 mL
pH 7 to 7.4
Content And Storage 4°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.