missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Vinyl Indoor Tape
MiniMark™ Vinyl Indoor Tape suitable for logistics identification.
Brand: Brady™ 710065
Code : 50
Additional Details : Weight : 0.40200kg
Description
- Indoor vinyl tape suitable for logistics identification.
Specifications
MiniMark Indoor Vinyl Tape | |
White | |
Roll | |
Thermal Transfer | |
Facility Identification, Floor marking, Hazard communication Identification, Pipe and valve marking, Safety Identification, Warehouse marking | |
1 | |
-40°C | |
1 Ea. | |
-40°C to +70°C | |
0.102 mm | |
115 ft. | |
1.967 in. | |
1.1 kg |
Permanent Acrylic, Permanent | |
Gloss | |
Vinyl | |
Labels | |
MiniMark Printer | |
2 Years | |
1 Roll/Box | |
-40°F to +158°F | |
0.004 in. | |
35 m x 50 mm | |
35 m | |
50 mm |