missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ MiniMark™ Pre-Printed Polypropylene Labels
Click to view available options
Height (Metric):
55 mm
75 mm
105 mm
Width (Metric):
85 mm
110 mm
155 mm
Includes:
1 Diamond
2 Diamonds, Vertical
3 Diamonds, Diamond Shape
3 Diamonds, Horizontal
4 Diamonds
4 Diamonds, Diamond Shape
4 Diamonds, Horizontal
Conditionnement:
Each
Description
- Supplied with pre-printed red diamonds.
- Ready for you to print the required warning.
- Compliant to the latest regulations.
Spécification
Spécification
| Certifications/Compliance | UL Recognised |
| Description | Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | Red on White |
| Finish | Gloss |
| Format | Die-cut |
| Material | Polypropylene |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Afficher plus |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu