missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Pre-Printed Polypropylene Labels

Product Code. 11739485
Change view
Click to view available options
Height (Metric):
55 mm
75 mm
105 mm
Width (Metric):
85 mm
110 mm
155 mm
Includes:
1 Diamond
2 Diamonds, Vertical
3 Diamonds, Diamond Shape
3 Diamonds, Horizontal
4 Diamonds
4 Diamonds, Diamond Shape
4 Diamonds, Horizontal
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Includes unitSize
11739485 55 mm 85 mm 3 Diamonds, Diamond Shape Each
11799465 105 mm 110 mm 1 Diamond Each
11709475 75 mm 110 mm 2 Diamonds, Vertical Each
11719475 75 mm 110 mm 3 Diamonds, Diamond Shape Each
11729475 75 mm 110 mm 3 Diamonds, Horizontal Each
11749475 75 mm 110 mm 4 Diamonds, Diamond Shape Each
11739475 75 mm 110 mm 4 Diamonds, Horizontal Each
11779455 105 mm 155 mm 1 Diamond Each
11789455 105 mm 155 mm 2 Diamonds, Vertical Each
11799455 105 mm 155 mm 3 Diamonds, Diamond Shape Each
11709465 105 mm 155 mm 3 Diamonds, Horizontal Each
11719465 105 mm 155 mm 4 Diamonds Each
11729485 55 mm 85 mm 2 Diamonds, Vertical Each
11719485 55 mm 85 mm 1 Diamond Each
11749485 55 mm 85 mm 3 Diamonds, Horizontal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11739485 Supplier Brady™ Supplier No. 710553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These MiniMark™ Pre-Printed Polypropylene Labels are supplied with pre-printed red diamonds and are ready for you to print the required warning.

  • Supplied with pre-printed red diamonds.
  • Ready for you to print the required warning.
  • Compliant to the latest regulations.

Specifications

Certifications/Compliance RoHS compliant using EU Directive 2002/95/EC
Description Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Gloss
Format Die-cut
Material Polypropylene
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) MiniMark Printer
Includes 3 Diamonds, Diamond Shape
Min. Operating Temperature (Metric) -20°C
No. per Pack 400 Labels/Box
Quantity 400 Pk.
Shape Diamond
Temperature Range (English) -4°F to +212°F
Temperature Range (Metric) -20°C to +100°C
Thickness (English) 0.0031 in.
Thickness (Metric) 0.079 mm
Dimensions (LxW) 55 x 85 mm
Length (English) 2.17 in.
Length (Metric) 55 mm
Width (English) 3.35 in.
Width (Metric) 85 mm
Height (English) 2.17 in.
Height (Metric) 55 mm
Alternate Part Number MNK-CLP-55x85-L3A-B7594-0.4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.