missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Pre-Printed Paper Labels

Product Code. 11799485
Click to view available options
Height (Metric):
55 mm
75 mm
105 mm
Width (Metric):
85 mm
110 mm
155 mm
Includes:
1 Diamond
2 Diamonds, Vertical
3 Diamonds, Diamond Shape
4 Diamonds, Diamond Shape
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11799485

Brand: Brady™ 710558

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

These MiniMark™ Pre-Printed Paper Labels are supplied with pre-printed red diamonds and are ready for you to print the required warning.

  • Supplied with pre-printed red diamonds.
  • Ready for you to print the required warning.
  • Compliant to the latest regulations.

Specifications

Certifications/Compliance RoHS, 2011/65/EU Directive
Description Pre-printed MiniMark Labels for CLP-GHS Hazardous Substances
Adhesive Type Acrylic, Permanent
Coating None
Color Red on White
Finish Matte
Format Die-cut
Material Paper
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification
For Use With (Equipment) MiniMark Printer
Includes 2 Diamonds, Vertical
Min. Operating Temperature (Metric) -40°C
No. per Pack 400 Labels/Box
Quantity 400 Pk.
Shape Diamond
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Dimensions (LxW) 55 x 85 mm
Length (English) 2.17 in.
Length (Metric) 55 mm
Width (English) 3.35 in.
Width (Metric) 85 mm
Height (English) 2.17 in.
Height (Metric) 55 mm
Alternate Part Number MNK-CLP-55x85-L2-B7610-0.4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.