missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark™ Ink Ribbons

Product Code. 11839514
Change view
Click to view available options
Color:
Black
Blue
Green
Red
White
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11839514 White Each
10475255 Black Each
11829514 Blue Each
11824125 Green Each
11864315 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11839514 Supplier Brady™ Supplier No. 710193
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These ribbons have standard scratch, smear and chemical resistance. MiniMark™ Ink Ribbons for B-7951 and B-7589 materials.

  • These ribbons have standard scratch, smear and chemical resistance.
  • MiniMark ribbon for B-7951 and B-7589 materials.

Specifications

Product Type Printer Ribbon
Color White
For Use With (Equipment) MiniMark Printer
Length (English) 98.4 yd.
Width (English) 4.33 in.
Length (Metric) 90 m
Width (Metric) 110 mm
Diameter (Metric) 12.7 mm I.D.
Dimensions (L x W) 90 m x 110 mm
Description 110 mm White Ribbon (1 Ea.)
Printable Sides Ink side out
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.