missing translation for 'onlineSavingsMsg'
Learn More

Brady™ MiniMark Indoor/Outdoor Vinyl Tape

Product Code. 10775674
Change view
Click to view available options
Color:
Green
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10775674 Yellow Each
10703424 Green Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10775674 Supplier Brady™ Supplier No. 120862
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tape constructed of B-7569 vinyl for indoor/outdoor use.

  • B-7569 is ideal for indoor/outdoor facility labels, including 5S labels, safety labels, warning labels, pipe markers, equipment labels and visual workplace labels

Specifications

Certifications/Compliance BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color Yellow
Display Blank
Finish Gloss
Format Continuous
Material Vinyl (B-7569)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Facility identification, Floor marking, Hazard communication identification, Pipe and valve marking, Safety identification, Warehouse marking
For Use With (Equipment) MiniMark
Labels per Row 1
Min. Operating Temperature (Metric) -20°C
No. per Pack 1 Roll/Box
Temperature Range (English) -4°F to +167°F
Temperature Range (Metric) -20°C to +75°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 33.53 m x 101.6 mm
Length (English) 110 ft.
Length (Metric) 33.53 m
Width (English) 4 in.
Width (Metric) 101.60 mm
Alternate Part Number TAPE CART MMK 100 MM YELLOW B7569 35M
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.