Learn More
Spectrum Labs™ MiniKross Sampler Fittings Kit
Click to view available options
:
For use with MiniKross TFF System
Description
- Includes a variety of fittings components
- Connectors
- Ties
- Gaskets
- Endcaps
- Clamps
- Reducers
Specifications
Specifications
| Connectors | TC, FLL |
| For Use With (Equipment) | MiniKross TFF System |
| Includes | 100 Black 6 in. Cable Ties10 1.5 in. Platinum Cured Silicone Sanitary Gaskets12 3/4 in. Platinum Cured Silicone Sanitary Gaskets6 1/2 in. Platinum Cured Silicone Sanitary Gaskets4 Clave Needle-Free Connectors4 3/4 in. TC Endcaps with Tab, Polypropylene4 3/4 in. OD Tubing Pinch Clamps4 1/5 in.-1/16 in. OD Tubing Pinch Clamps8 1/2-3/4 in. Plastic TC Clamps4 1.5 in. Plastic TC Clamps6 1/2-3/4 in. TC x 1/4 in. HB Adapters, Polyproylene6 1/2-3/4 in. TC x 3/8 in. HB Adapters, Polyproylene2 3/4 in. TC x 1/4 in. HB Adapters x Female Luer Lock Pro-Connex2 3/4 in. TC x 3/8 in. HB Adapters x Female Luer Lock Pro-Connex4 Male Luer Caps, Polypropylene4 1.5 in. to 1/2-3/4 in. Conical Reducers |