missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ MiniFix Micropipettor

Product Code. 10257222
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
500 μL
Compatible Tips:
Kartell™ 956/959
Kartell™ 960/958
Kartell™ 961
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Compatible Tips unitSize
10257222 20 μL Kartell™ 960/958 Each
10798293 100 μL Kartell™ 960/958 Each
10181473 500 μL Kartell™ 961 Each
10790404 10 μL Kartell™ 960/958 Each
10053702 25 μL Kartell™ 960/958 Each
10434242 5 μL Kartell™ 956/959 Each
10389142 50 μL Kartell™ 960/958 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10257222 Supplier Kartell™ Supplier No. 0128700
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An innovative product line of fixed-volume air dispacement mini-micropipettes. Kartell™ MiniFix Micropipettor is designed for fast, repetitive, and precise operation and are competitively priced.

  • Fully autoclavable
  • Designed for fast, repetitive and precise operation
  • Very compact in terms of size and weight
  • Designed for routine or biological hazard work
  • Kartell™ tips fit on the conical shaft as well as most other commercially available tips
  • The plunger button has a different color based on dosing capacity
  • Individually calibrated in compliance with ISO8655, the new international standard for piston-operated volumetric apparatus
  • Carries the CE conformity mark

Specifications

Accuracy ±0.5%
Certifications/Compliance CE/IVD
Compatible Tips Kartell™ 960/958
Autoclavable Autoclavable
Color Red
For Use With in Situ Analyses and Sampling, Routine or Biological Hazard Work
Model MiniFix
Number of Channels 1
Type Micropipette
Volume (Metric) 20 μL
For Use With (Application) Routine or biological hazard work, in situ analyses and sampling
Quantity 1/Pk.
Length (Metric) 13 cm
Weight (Metric) 10 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.