Learn More
Specac™ Minidiff Plus Diffuse Reflectance Accessory for FT-IR Spectroscopy
Product Code. 15270008
Encompass_Preferred
Change view
Click to view available options
Type:
Minidiff Plus Diffuse Reflectance Accessory
Product Code. Type
15270008 Minidiff Plus Diffuse Reflectance Accessory
1 options
Product Code. 15270008 Supplier Specac™ Supplier No. GS04510
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Diffuse Reflectance Accessory for routine analysis. High performance solids analysis is made simple through a minimal alignment optical system. Solid sampling versatility is maintained through use of sampling cups and Diabrase abrasive sampling cups. Suitable for powders, solids and liquids.
Specifications
Type Minidiff Plus Diffuse Reflectance Accessory
For Use With (Application) FT-IR Spectroscopy