Learn More
Greiner Bio-One MiniCollect™ Tube. Lavender Cap, K3E K3EDTA, Non-Sterile
Product Code. 15758169
Change view
Click to view available options
Capacity (Metric):
0.25, 0.5 mL
1 mL
Unit Size:
Pack of 2000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15758169 1 mL Pack of 500
15748169 0.25, 0.5 mL Pack of 500
15830146 0.25, 0.5 mL Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15758169 Supplier Greiner Bio-One Supplier No. 450531
 Restricted Item
In Stock
Add to basket
Add to basket
Inner wall is coated with anticoagulant that binds calcium ions and interrupts clotting.
  • Inner wall is coated with spray-dried K3E K3EDTA as an anticoagulant to bind calcium ions and interrupt the clotting cascade.
  • Used for hematology.
  • Available in different quantities.
TRUSTED_SUSTAINABILITY
Specifications
Additive K3 E K3 EDTA
Sterility Non-sterile
Capacity (Metric) 1 mL
Closure Included Yes
Closure Color Lavender