Learn More
IBA Lifesciences Mini Spin Columns
Shop All IBA Lifesciences Products
Click to view available options
Kit Contents:
10 columns
50 columns
Unit Size:
Pack of 10
Pack of 50
Description
- Mini Spin Columns are the ideal solution to treat small volumes in combination with Selector Resins.
- They fit into every microcentrifuge and are compatible with all standard 1.5 ml or 2.0 ml tubes.
- Mini Spin Columns are provided with inserted 10 μm filters, screw caps, and snap-off plugs
Specifications
Specifications
| Kit Contents | 50 columns |
| For Use With (Application) | Rapid Purification of Recombinant Proteins |