Learn More
IBA Lifesciences Mini Spin Columns
Product Code. 17257590 Shop All IBA Lifesciences Products
Click to view available options
Kit Contents:
10 columns
50 columns
Unit Size:
Pack of 10
Pack of 50
Product Code. 17257590
Brand: IBA Lifesciences 29102010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For rapid purification of recombinant proteins
  • Mini Spin Columns are the ideal solution to treat small volumes in combination with Selector Resins.
  • They fit into every microcentrifuge and are compatible with all standard 1.5 ml or 2.0 ml tubes.
  • Mini Spin Columns are provided with inserted 10 μm filters, screw caps, and snap-off plugs
Specifications
Kit Contents 10 columns
For Use With (Application) Rapid Purification of Recombinant Proteins