missing translation for 'onlineSavingsMsg'
Learn More

Scie Plas Mini-gel Gel Electrophoresis Unit, Battery Operated

Product Code. 15835541
Click to view available options
Gel Dimensions:
(W x L): 6 x 7.5 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15835541

Brand: Scie Plas HBSET

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Allows direct gel casting in tank without additional accessories.

  • Complete system
  • Utilizes low voltage
  • Integrated safety lid to ensure gel remains inaccessible during use
  • UV transparent acrylic - for direct visualisation of gel within tank
  • Ultra-compact tank, minimises buffer volume required to cover gel
  • Small size to allow safe, easy transportation from bench to transilluminator
  • Built-in plastic casting gates allow direct gel casting in tank without additional costly accessories
  • Compact size: 170mm x 110mm x 35mm (D x W x H)
  • Gel dimensions: 75mm x 60mm (D x W)
  • Maximum number of samples: 32
  • Built-in gel casting tray

Specifications

Type Gel Electrophoresis Unit
Gel Dimensions (W x L): 6 x 7.5 cm
Sample Size 32
Volume (Metric) Buffer 50 mL
Voltage 18 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.