Learn More
Specac™ Mini-Film Maker Kit for IR Transmission Analysis
Product Code. 15200008
Encompass_Preferred
Click to view available options
Type:
Mini-Film Maker Kit
Product Code. 15200008
Brand: Specac™ GS03970
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A dedicated polymer thin film-making accessory for IR transmission analysis. This kit is designed to produce high quality thin films of polymer and plastic materials for IR Transmission analysis. Capable of operating up to 250°C through use of Temperature Controller.
Specifications
Type Mini-Film Maker Kit
Description Capable of operating up to 250°C through use of Temperature Controller.