missing translation for 'onlineSavingsMsg'
Learn More

Koettermann™ Mini-Cabinet for Toxic Products

Product Code. 10528862
Click to view available options
No. of Doors:
1
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10528862

Brand: Koettermann™ 379005

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Koettermann™ Mini-Cabinet for Toxic Products is a small, lockable, non-ventilated cabinet for storing poisonous materials.

  • Small size
  • Not ventilated
  • For placement on a table or inside another cabinet
  • Lockable
  • Includes fixed shelf
  • Includes toxic material signage

Specifications

Dimensions (D x W x H) 35 x 32 x 37 cm
For Use With (Application) Storage of toxic chemicals
Includes Fixed shelf, lock
No. of Doors 1
Color Gray
Material Steel (Galvanized)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.