Learn More
SPEX CertiPrep™ Minerals Testing Kit
Minerals Testing Kit, 1,000μg/mL, for ICP-MS
Brand: SPEX CertiPrep™ SPXMT-KIT
Additional Details : CAS Number : 471-34-1 Weight : 0.03000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
Claritas PPT Grade Minerals Testing Kit for ICP-MS, 1,000μg/mL (1,000 ppm), 6 each 30mLKit contains:
- Calcium, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Iron, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Potassium, 1,000μg/mL, for ICP-MS, 30mL, Matrix: 2% HNO3
- Magnesium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
- Sodium, 1,000μg/mL, for ICP-MS, 30mL Matrix: 2% HNO3
Specifications
2% HNO3 | |
Mineral teesting | |
30 mL | |
Ambient |