missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mineralocorticoid R/NR3C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | Mineralocorticoid R/NR3C2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Simple Western, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, Simple Western, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626805
|
Novus Biologicals
NBP2-99010-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674905
|
Novus Biologicals
NBP2-99010-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mineralocorticoid R/NR3C2 Polyclonal antibody specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Mineralocorticoid R/NR3C2 | |
| Western Blot, Simple Western, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Mineralocorticoid R/NR3C2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Simple Western, Immunocytochemistry/Immunofluorescence 1:300-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| 4306 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title