missing translation for 'onlineSavingsMsg'
Learn More

Mineralocorticoid R/NR3C2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18396302 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396302 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396302 Supplier Novus Biologicals Supplier No. NBP299010AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Mineralocorticoid R/NR3C2 Polyclonal specifically detects Mineralocorticoid R/NR3C2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mineralocorticoid R/NR3C2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias FLJ41052, MGC133092, mineralocorticoid receptor 1, mineralocorticoid receptor delta, MLRmineralocorticoid receptor, MRMCRaldosterone receptor, NR3C2VIT, Nuclear receptor subfamily 3 group C member 2, nuclear receptor subfamily 3, group C, member 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Mineralocorticoid R/NR3C2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4306
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.